Product Details

SNP ID
rs143076468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133264602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCAGCCAGGGGAAGCCCGTGAG[A/G]GTGAAGAGTGAGTGACTTCCCTCCT
Phenotype
MIM: 602267
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM8 PubMed Links

Gene Details

Gene
ADAM8
Gene Name
ADAM metallopeptidase domain 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109.4 Intron NP_001100.3
NM_001164489.1 Intron NP_001157961.1
NM_001164490.1 Intron NP_001157962.1
XM_011539117.2 Intron XP_011537419.2
XM_017015465.1 Intron XP_016870954.1
XM_017015466.1 Intron XP_016870955.1

View Full Product Details