Product Details

SNP ID
rs145218772
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:123149966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCCAACATGGCGCACGCGTCGG[C/G]GCAGACTCTGGTGAGCATGCCGCTG
Phenotype
MIM: 603719 MIM: 600647
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BUB3 PubMed Links

Gene Details

Gene
BUB3
Gene Name
BUB3, mitotic checkpoint protein
There are no transcripts associated with this gene.

Gene
HMX2
Gene Name
H6 family homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005519.1 922 Missense Mutation GCG,GGG A222G NP_005510.1
XM_005269743.3 922 Missense Mutation GCG,GGG A222G XP_005269800.1

View Full Product Details