Product Details

SNP ID
rs147635148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:15049666 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACATTTTCAACTGCTTATACAAA[A/G]ACCCTGAGGCAACTTTTAAGCTGAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OLAH PubMed Links

Gene Details

Gene
OLAH
Gene Name
oleoyl-ACP hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039702.2 282 Missense Mutation AAC,GAC N22D NP_001034791.1
NM_018324.2 282 Missense Mutation AAC,GAC N22D NP_060794.1
XM_017016376.1 282 Missense Mutation AAC,GAC N22D XP_016871865.1

View Full Product Details