Product Details
- SNP ID
-
rs148583159
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:30845284 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTGCCTTTTCAGTCTTGGACATG[C/T]ATGTGATTCCTCGGAGGAGGAATGA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF438
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3740236] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF438
- Gene Name
- zinc finger protein 438
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001143766.1 |
2639 |
Missense Mutation |
ACA,GCA |
T722A |
NP_001137238.1 |
NM_001143767.1 |
2639 |
Missense Mutation |
ACA,GCA |
T722A |
NP_001137239.1 |
NM_001143768.1 |
2639 |
Missense Mutation |
ACA,GCA |
T722A |
NP_001137240.1 |
NM_001143769.1 |
2639 |
Missense Mutation |
ACA,GCA |
T673A |
NP_001137241.1 |
NM_001143770.1 |
2639 |
Missense Mutation |
ACA,GCA |
T712A |
NP_001137242.1 |
NM_001143771.1 |
2639 |
Missense Mutation |
ACA,GCA |
T712A |
NP_001137243.1 |
NM_182755.2 |
2639 |
Missense Mutation |
ACA,GCA |
T722A |
NP_877432.1 |
XM_006717393.3 |
2639 |
Missense Mutation |
ACA,GCA |
T722A |
XP_006717456.1 |
XM_006717396.3 |
2639 |
Missense Mutation |
ACA,GCA |
T722A |
XP_006717459.1 |
XM_006717398.3 |
2639 |
Missense Mutation |
ACA,GCA |
T712A |
XP_006717461.1 |
XM_006717399.3 |
2639 |
Missense Mutation |
ACA,GCA |
T712A |
XP_006717462.1 |
XM_011519371.2 |
2639 |
Missense Mutation |
ACA,GCA |
T722A |
XP_011517673.1 |
XM_011519372.2 |
2639 |
Missense Mutation |
ACA,GCA |
T722A |
XP_011517674.1 |
XM_011519373.2 |
2639 |
Missense Mutation |
ACA,GCA |
T722A |
XP_011517675.1 |
XM_011519374.2 |
2639 |
Missense Mutation |
ACA,GCA |
T722A |
XP_011517676.1 |
XM_011519376.2 |
2639 |
Missense Mutation |
ACA,GCA |
T712A |
XP_011517678.1 |
XM_011519377.2 |
2639 |
Missense Mutation |
ACA,GCA |
T712A |
XP_011517679.1 |
XM_017015863.1 |
2639 |
Missense Mutation |
ACA,GCA |
T733A |
XP_016871352.1 |
XM_017015864.1 |
2639 |
Missense Mutation |
ACA,GCA |
T722A |
XP_016871353.1 |
XM_017015865.1 |
2639 |
Missense Mutation |
ACA,GCA |
T722A |
XP_016871354.1 |
XM_017015866.1 |
2639 |
Missense Mutation |
ACA,GCA |
T722A |
XP_016871355.1 |
XM_017015867.1 |
2639 |
Missense Mutation |
ACA,GCA |
T722A |
XP_016871356.1 |
XM_017015868.1 |
2639 |
Missense Mutation |
ACA,GCA |
T722A |
XP_016871357.1 |
XM_017015869.1 |
2639 |
Missense Mutation |
ACA,GCA |
T722A |
XP_016871358.1 |
XM_017015870.1 |
2639 |
Missense Mutation |
ACA,GCA |
T722A |
XP_016871359.1 |
XM_017015871.1 |
2639 |
Missense Mutation |
ACA,GCA |
T722A |
XP_016871360.1 |
XM_017015872.1 |
2639 |
Missense Mutation |
ACA,GCA |
T722A |
XP_016871361.1 |
XM_017015873.1 |
2639 |
Missense Mutation |
ACA,GCA |
T712A |
XP_016871362.1 |
XM_017015874.1 |
2639 |
Missense Mutation |
ACA,GCA |
T712A |
XP_016871363.1 |
XM_017015875.1 |
2639 |
Missense Mutation |
ACA,GCA |
T286A |
XP_016871364.1 |
XM_017015876.1 |
2639 |
Missense Mutation |
ACA,GCA |
T286A |
XP_016871365.1 |
XM_017015877.1 |
2639 |
Missense Mutation |
ACA,GCA |
T286A |
XP_016871366.1 |
XM_017015878.1 |
2639 |
Missense Mutation |
ACA,GCA |
T286A |
XP_016871367.1 |
XM_017015879.1 |
2639 |
Missense Mutation |
ACA,GCA |
T286A |
XP_016871368.1 |
XM_017015880.1 |
2639 |
Missense Mutation |
ACA,GCA |
T286A |
XP_016871369.1 |
XM_017015881.1 |
2639 |
Missense Mutation |
ACA,GCA |
T286A |
XP_016871370.1 |
XM_017015882.1 |
2639 |
Missense Mutation |
ACA,GCA |
T286A |
XP_016871371.1 |
View Full Product Details