Product Details
- SNP ID
-
rs149570877
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:992565 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTCATAAACATTACCAAATACATC[G/T]CATTAGACATTTTTACATGAGAAAA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GTPBP4
PubMed Links
Gene Details
- Gene
- GTPBP4
- Gene Name
- GTP binding protein 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_012341.2 |
196 |
Missense Mutation |
CGC,CTC |
R42L |
NP_036473.2 |
View Full Product Details