Product Details

SNP ID
rs111240465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64243835 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGACTGTTTCTTTGTGCATCTT[C/T]AACAGGGTATGGAGAGCGGGGAGCT
Phenotype
MIM: 604189 MIM: 186946 MIM: 600230 MIM: 601140 MIM: 601398
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC4 PubMed Links

Gene Details

Gene
DNAJC4
Gene Name
DnaJ heat shock protein family (Hsp40) member C4
There are no transcripts associated with this gene.

Gene
FKBP2
Gene Name
FK506 binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135208.1 Intron NP_001128680.1
NM_004470.3 Intron NP_004461.2
NM_057092.2 Intron NP_476433.1
XM_005273848.3 Intron XP_005273905.1
Gene
LOC105369340
Gene Name
uncharacterized LOC105369340
There are no transcripts associated with this gene.

Gene
PLCB3
Gene Name
phospholipase C beta 3
There are no transcripts associated with this gene.

Gene
PPP1R14B
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 14B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138689.2 Intron NP_619634.1
Gene
VEGFB
Gene Name
vascular endothelial growth factor B
There are no transcripts associated with this gene.

View Full Product Details