Product Details

SNP ID
rs114106353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:92233434 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTGCTGGGTTCTTCAGGCCCGT[A/G]TCATTGTTGGCAAAGTATACTAAGA
Phenotype
MIM: 612483
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAT3 PubMed Links

Gene Details

Gene
FAT3
Gene Name
FAT atypical cadherin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008781.2 Intron NP_001008781.2
XM_017017178.1 Intron XP_016872667.1
XM_017017179.1 Intron XP_016872668.1
XM_017017180.1 Intron XP_016872669.1
XM_017017181.1 Intron XP_016872670.1
XM_017017182.1 Intron XP_016872671.1
XM_017017183.1 Intron XP_016872672.1
XM_017017184.1 Intron XP_016872673.1
XM_017017185.1 Intron XP_016872674.1
XM_017017186.1 Intron XP_016872675.1
XM_017017187.1 Intron XP_016872676.1

View Full Product Details