Product Details
- SNP ID
-
rs138363552
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:124424478 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGACCCTTGGGGACATGGTGACCA[A/T]GTAGAGCAGGGGGTTGCAGATGGCC
- Phenotype
-
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR8B4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4057750] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR8B4
- Gene Name
- olfactory receptor family 8 subfamily B member 4 (gene/pseudogene)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001005196.1 |
394 |
Missense Mutation |
ATG,TTG |
M132L |
NP_001005196.1 |
View Full Product Details