Product Details

SNP ID
rs138732188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:106008244 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGCTGAAGGAAAATCATCAGGT[C/T]AAGCAAGCCCTTGGTCTCAGAATAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MSANTD4 PubMed Links

Gene Details

Gene
MSANTD4
Gene Name
Myb/SANT DNA binding domain containing 4 with coiled-coils
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318747.1 2931 UTR 3 NP_001305676.1
NM_001318748.1 2931 UTR 3 NP_001305677.1
NM_001318749.1 2931 UTR 3 NP_001305678.1
NM_001318750.1 2931 UTR 3 NP_001305679.1
NM_032424.2 2931 UTR 3 NP_115800.1
XM_011543022.2 2931 UTR 3 XP_011541324.1
XM_017018416.1 2931 UTR 3 XP_016873905.1
XM_017018417.1 2931 UTR 3 XP_016873906.1

View Full Product Details