Product Details

SNP ID
rs138881906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119190787 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCCAGGGCCTGGTCCCGCTCTC[C/T]GAGAGCTTGTTCAGCCTCTTCCCGG
Phenotype
MIM: 611947 MIM: 607146
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC153 PubMed Links

Gene Details

Gene
CCDC153
Gene Name
coiled-coil domain containing 153
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145018.1 708 Missense Mutation NP_001138490.1
XM_006718818.3 708 Nonsense Mutation XP_006718881.1
XM_011542763.2 708 Nonsense Mutation XP_011541065.1
XM_011542764.2 708 Nonsense Mutation XP_011541066.1
XM_011542770.2 708 Nonsense Mutation XP_011541072.1
XM_011542771.2 708 Nonsense Mutation XP_011541073.1
XM_011542772.2 708 Nonsense Mutation XP_011541074.1
XM_011542774.2 708 Nonsense Mutation XP_011541076.1
XM_017017577.1 708 Nonsense Mutation XP_016873066.1
XM_017017578.1 708 UTR 3 XP_016873067.1
XM_017017579.1 708 Nonsense Mutation XP_016873068.1
XM_017017580.1 708 Missense Mutation XP_016873069.1
Gene
NLRX1
Gene Name
NLR family member X1
There are no transcripts associated with this gene.

Gene
PDZD3
Gene Name
PDZ domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168468.1 708 Intron NP_001161940.1
NM_024791.3 708 Intron NP_079067.3
XM_011543000.2 708 Intron XP_011541302.1
XM_011543001.2 708 Intron XP_011541303.1
XM_011543002.2 708 Intron XP_011541304.1
XM_011543005.2 708 Intron XP_011541307.1
XM_017018334.1 708 Intron XP_016873823.1
XM_017018335.1 708 Intron XP_016873824.1
XM_017018336.1 708 Intron XP_016873825.1
XM_017018337.1 708 Intron XP_016873826.1

View Full Product Details