Product Details

SNP ID
rs139614081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123943181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTGAGAACAGTGGCAAGAATGC[A/G]GGCATAGGAAACTGAGGTCAGAGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4D5 PubMed Links

Gene Details

Gene
OR4D5
Gene Name
olfactory receptor family 4 subfamily D member 5
There are no transcripts associated with this gene.

Gene
OR6T1
Gene Name
olfactory receptor family 6 subfamily T member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005187.1 573 Missense Mutation CGC,TGC R220C NP_001005187.1
XM_017017325.1 573 Missense Mutation CGC,TGC R162C XP_016872814.1

View Full Product Details