Product Details

SNP ID
rs139874965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67393563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGTGGAAAAATGCTGCATCTCCC[G/T]GAATGGCCGGAGCAGCCGCCTGGTG
Phenotype
MIM: 176875 MIM: 603761
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC100130987 PubMed Links

Gene Details

Gene
LOC100130987
Gene Name
uncharacterized LOC100130987
There are no transcripts associated with this gene.

Gene
PPP1CA
Gene Name
protein phosphatase 1 catalytic subunit alpha
There are no transcripts associated with this gene.

Gene
RAD9A
Gene Name
RAD9 checkpoint clamp component A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243224.1 607 Missense Mutation CGG,CTG R25L NP_001230153.1
NM_004584.2 607 Missense Mutation CGG,CTG R101L NP_004575.1
XM_006718652.3 607 Missense Mutation CGG,CTG R112L XP_006718715.1
XM_017018097.1 607 UTR 5 XP_016873586.1
XM_017018098.1 607 UTR 5 XP_016873587.1

View Full Product Details