Product Details

SNP ID
rs139900371
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:101891438 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTAGATTTGCTAGACCACACTC[A/G]TTAAACCACCAGCCGGTCTTGTTAT
Phenotype
MIM: 607666
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANGPTL5 PubMed Links

Gene Details

Gene
ANGPTL5
Gene Name
angiopoietin like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178127.4 1178 Silent Mutation AAA,AAG K336K NP_835228.2
XM_011542735.2 1178 Silent Mutation AAA,AAG K271K XP_011541037.1
XM_017017466.1 1178 Silent Mutation AAA,AAG K196K XP_016872955.1

View Full Product Details