Product Details

SNP ID
rs140608295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:133842097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTGAGCTGAGACATCTGTAAGA[C/T]GGGATATGCTGCAGGGGACAGAGGA
Phenotype
MIM: 609805
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPATA19 PubMed Links

Gene Details

Gene
SPATA19
Gene Name
spermatogenesis associated 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291992.1 538 Missense Mutation CAT,CGT H149R NP_001278921.1
NM_174927.2 538 Missense Mutation CAT,CGT H149R NP_777587.1

View Full Product Details