Product Details

SNP ID
rs140762238
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63813701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGCCGCACCATGGCCCTAAAGGC[C/G]GAGGGCGCCGCACTCGACTGCTTCG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C11orf84 PubMed Links

Gene Details

Gene
C11orf84
Gene Name
chromosome 11 open reading frame 84
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138471.2 328 Silent Mutation GCC,GCG A5A NP_612480.1
XM_005273782.2 328 Silent Mutation GCC,GCG A5A XP_005273839.1
XM_005273783.2 328 Silent Mutation GCC,GCG A5A XP_005273840.1
XM_006718437.1 328 Silent Mutation GCC,GCG A5A XP_006718500.1
XM_011544770.1 328 Silent Mutation GCC,GCG A5A XP_011543072.1
XM_011544771.1 328 Silent Mutation GCC,GCG A5A XP_011543073.1
XM_011544772.1 328 Silent Mutation GCC,GCG A5A XP_011543074.1

View Full Product Details