Product Details

SNP ID
rs141662559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:106079518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGCAGAGAAATTGAATATCCCT[G/T]GGAATCATATTCGTTTGCAAAGAAC
Phenotype
MIM: 607756
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AASDHPPT PubMed Links

Gene Details

Gene
AASDHPPT
Gene Name
aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015423.2 381 Missense Mutation GGG,TGG G79W NP_056238.2
Gene
KBTBD3
Gene Name
kelch repeat and BTB domain containing 3
There are no transcripts associated with this gene.

View Full Product Details