Product Details

SNP ID
rs141695803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74457254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCACCGTCCCAGCCCTCTCGTGTT[A/G]GATCATAGACACACGGTTCTTGATA
Phenotype
MIM: 604433
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNE3 PubMed Links

Gene Details

Gene
KCNE3
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 3
There are no transcripts associated with this gene.

View Full Product Details