Product Details

SNP ID
rs141795168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:298678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTTTGGAGCCAAAACGCCGGGC[C/T]GCTTCCAGGTCACCAACCACCTTCT
Phenotype
MIM: 605578 MIM: 614757 MIM: 617032
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFITM2 PubMed Links

Gene Details

Gene
IFITM2
Gene Name
interferon induced transmembrane protein 2
There are no transcripts associated with this gene.

Gene
IFITM5
Gene Name
interferon induced transmembrane protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025295.2 258 Silent Mutation GCA,GCG A74A NP_001020466.1
Gene
PGGHG
Gene Name
protein-glucosylgalactosylhydroxylysine glucosidase
There are no transcripts associated with this gene.

View Full Product Details