Product Details

SNP ID
rs142613980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:13492626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCCATCGAGTTCAGATGTTTTC[C/G]CAGGTTATGCATAAGCTGTATTTCA
Phenotype
MIM: 168450
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PTH PubMed Links

Gene Details

Gene
PTH
Gene Name
parathyroid hormone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000315.3 266 Missense Mutation CGA,GGA R43G NP_000306.1
NM_001316352.1 266 Missense Mutation CGA,GGA R75G NP_001303281.1

View Full Product Details