Product Details

SNP ID
rs143262863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56159552 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATTGGGCTGCATGTACATGAAGA[A/G]TAGAGACCCATAGAACACAACCACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR8K5 PubMed Links

Gene Details

Gene
OR8K5
Gene Name
olfactory receptor family 8 subfamily K member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004058.2 766 Missense Mutation CTC,TTC L256F NP_001004058.2

View Full Product Details