Product Details

SNP ID
rs143558098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66278519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGGTGCTGTGCGCCTCCCTCAT[C/T]TTCTTTGTCATCTGGCACGTAAGGC
Phenotype
MIM: 611288 MIM: 612565 MIM: 611484
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNIH2 PubMed Links

Gene Details

Gene
CNIH2
Gene Name
cornichon family AMPA receptor auxiliary protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182553.2 319 Silent Mutation ATC,ATT I21I NP_872359.1
Gene
RAB1B
Gene Name
RAB1B, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
YIF1A
Gene Name
Yip1 interacting factor homolog A, membrane trafficking protein
There are no transcripts associated with this gene.

View Full Product Details