Product Details

SNP ID
rs143747626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55572462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTTGGCTGCCTGGAGATCTTCA[A/T]CCTCATCCTCACGGCTGTTGACCGC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR4C16 PubMed Links

Gene Details

Gene
OR4C16
Gene Name
olfactory receptor family 4 subfamily C member 16 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004701.2 335 Missense Mutation AAC,ATC N112I NP_001004701.2

View Full Product Details