Product Details

SNP ID
rs143788676
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65321037 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACACCATTCATATTGGCAGTGGC[A/G]GCGGCAGTGACATGTTTGGCGACAT
Phenotype
MIM: 606132
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC42EP2 PubMed Links

Gene Details

Gene
CDC42EP2
Gene Name
CDC42 effector protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006779.3 742 Missense Mutation AGC,GGC S47G NP_006770.1
XM_017017094.1 742 Missense Mutation AGC,GGC S47G XP_016872583.1

View Full Product Details