Product Details

SNP ID
rs143916090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119193163 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGAAACCTACCTAGCTGCCCCC[A/G]AAGGCCTTTGACTTCTTCCTCCAGC
Phenotype
MIM: 611947 MIM: 607146
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC153 PubMed Links

Gene Details

Gene
CCDC153
Gene Name
coiled-coil domain containing 153
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145018.1 642 Missense Mutation CGG,TGG R113W NP_001138490.1
XM_006718818.3 642 Missense Mutation CGG,TGG R85W XP_006718881.1
XM_011542763.2 642 Missense Mutation CGG,TGG R273W XP_011541065.1
XM_011542764.2 642 Missense Mutation CGG,TGG R273W XP_011541066.1
XM_011542770.2 642 Missense Mutation CGG,TGG R113W XP_011541072.1
XM_011542771.2 642 Missense Mutation CGG,TGG R105W XP_011541073.1
XM_011542772.2 642 Missense Mutation CGG,TGG R105W XP_011541074.1
XM_011542774.2 642 Missense Mutation CGG,TGG R175W XP_011541076.1
XM_017017577.1 642 Missense Mutation CGG,TGG R201W XP_016873066.1
XM_017017578.1 642 Missense Mutation CGG,TGG R148W XP_016873067.1
XM_017017579.1 642 Missense Mutation CGG,TGG R42W XP_016873068.1
XM_017017580.1 642 Missense Mutation CGG,TGG R113W XP_016873069.1
Gene
NLRX1
Gene Name
NLR family member X1
There are no transcripts associated with this gene.

Gene
PDZD3
Gene Name
PDZ domain containing 3
There are no transcripts associated with this gene.

View Full Product Details