Product Details

SNP ID
rs145052992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60700960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCCAGGAATTATGTCTCACGTG[A/C]CCCTGTATCCAAACAGCCAGCCGCA
Phenotype
MIM: 606549
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MS4A8 PubMed Links

Gene Details

Gene
MS4A8
Gene Name
membrane spanning 4-domains A8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031457.1 303 Missense Mutation ACC,CCC T34P NP_113645.1

View Full Product Details