Product Details
- SNP ID
-
rs146148230
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:116749707 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGCGTAGGCCATGGTAAAAAGCT[C/T]AGATTTCATCCAGAAACGATGGGTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BUD13
PubMed Links
Gene Details
- Gene
- BUD13
- Gene Name
- BUD13 homolog
View Full Product Details