Product Details

SNP ID
rs148535951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68896600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTCAGAGGTCACCTTCCACTGG[C/T]GGCTGGCAAAGTGCACCACGGCAAA
Phenotype
MIM: 600502 MIM: 611834
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGHMBP2 PubMed Links

Gene Details

Gene
IGHMBP2
Gene Name
immunoglobulin mu binding protein 2
There are no transcripts associated with this gene.

Gene
MRPL21
Gene Name
mitochondrial ribosomal protein L21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181514.1 349 Missense Mutation CAC,CGC H104R NP_852615.1
NM_181515.1 349 Missense Mutation CAC,CGC H19R NP_852616.1
XM_005273823.4 349 Missense Mutation CAC,CGC H104R XP_005273880.1

View Full Product Details