Product Details

SNP ID
rs148924410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65999358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGCTTGTTCCTCACCTGTTCCT[A/G]TTGTTGTGTTTCTCAGCCACTTCAG
Phenotype
MIM: 603811
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BANF1 PubMed Links

Gene Details

Gene
BANF1
Gene Name
barrier to autointegration factor 1
There are no transcripts associated with this gene.

Gene
EIF1AD
Gene Name
eukaryotic translation initiation factor 1A domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242481.1 489 Silent Mutation AAC,AAT N115N NP_001229410.1
NM_001242482.1 489 Silent Mutation AAC,AAT N115N NP_001229411.1
NM_001242483.1 489 Silent Mutation AAC,AAT N115N NP_001229412.1
NM_001242484.1 489 Silent Mutation AAC,AAT N115N NP_001229413.1
NM_001242485.1 489 Silent Mutation AAC,AAT N115N NP_001229414.1
NM_001242486.1 489 Silent Mutation AAC,AAT N115N NP_001229415.1
NM_032325.3 489 Silent Mutation AAC,AAT N115N NP_115701.2
XM_017018412.1 489 Silent Mutation AAC,AAT N115N XP_016873901.1

View Full Product Details