Product Details
- SNP ID
-
rs149941214
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:75437023 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCAGCATGATCTGCTCAGGGTTGT[A/G]GCTCCGTATGCCACCCAGGCGCCAC
- Phenotype
-
MIM: 609632
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GDPD5
PubMed Links
Gene Details
- Gene
- GDPD5
- Gene Name
- glycerophosphodiester phosphodiesterase domain containing 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_030792.6 |
2851 |
Missense Mutation |
CAC,TAC |
H528Y |
NP_110419.5 |
XM_006718697.3 |
2851 |
Missense Mutation |
CAC,TAC |
H528Y |
XP_006718760.1 |
XM_011545276.2 |
2851 |
Missense Mutation |
CAC,TAC |
H528Y |
XP_011543578.1 |
XM_011545277.2 |
2851 |
Missense Mutation |
CAC,TAC |
H528Y |
XP_011543579.1 |
XM_011545278.2 |
2851 |
Missense Mutation |
CAC,TAC |
H528Y |
XP_011543580.1 |
XM_011545279.2 |
2851 |
Missense Mutation |
CAC,TAC |
H528Y |
XP_011543581.1 |
XM_011545280.1 |
2851 |
Missense Mutation |
CAC,TAC |
H471Y |
XP_011543582.1 |
XM_011545281.2 |
2851 |
Missense Mutation |
CAC,TAC |
H390Y |
XP_011543583.1 |
XM_011545285.2 |
2851 |
Missense Mutation |
CAC,TAC |
H390Y |
XP_011543587.1 |
XM_011545286.1 |
2851 |
Missense Mutation |
CAC,TAC |
H283Y |
XP_011543588.1 |
XM_011545287.1 |
2851 |
Missense Mutation |
CAC,TAC |
H283Y |
XP_011543589.1 |
- Gene
- KLHL35
- Gene Name
- kelch like family member 35
There are no transcripts associated with this gene.
View Full Product Details