Product Details

SNP ID
rs150004398
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63815418 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTTTCCGAGATGGGGAAGGCTG[C/T]GGTGCTCTTGGGAGACTGGGAGTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf84 PubMed Links

Gene Details

Gene
C11orf84
Gene Name
chromosome 11 open reading frame 84
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138471.2 Intron NP_612480.1
XM_005273782.2 Intron XP_005273839.1
XM_005273783.2 Intron XP_005273840.1
XM_006718437.1 Intron XP_006718500.1
XM_011544770.1 Intron XP_011543072.1
XM_011544771.1 Intron XP_011543073.1
XM_011544772.1 Intron XP_011543074.1

View Full Product Details