Product Details
- SNP ID
-
rs151023546
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:118956953 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGAGAGCCTGCTGGTTGCCTTGCC[C/T]CCCTGTCACCTCACAGGAGGCAATG
- Phenotype
-
MIM: 611558
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
UPK2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3886020] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- UPK2
- Gene Name
- uroplakin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006760.3 |
200 |
Silent Mutation |
CCC,CCT |
P49P |
NP_006751.1 |
View Full Product Details