Product Details

SNP ID
rs151168710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18021110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTCTTTCAGGATCTGAATACTCC[A/G]TGTATATGGATTATACTTCACTCCA
Phenotype
MIM: 606051 MIM: 191060
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERGEF PubMed Links

Gene Details

Gene
SERGEF
Gene Name
secretion regulating guanine nucleotide exchange factor
There are no transcripts associated with this gene.

Gene
TPH1
Gene Name
tryptophan hydroxylase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004179.2 1242 Missense Mutation CGG,TGG R406W NP_004170.1

View Full Product Details