Product Details
- SNP ID
-
rs138455492
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:54633211 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCTTCCTGGGCAGGATCAGCACC[C/T]GTCGAGTCAGAGGAGGCATCTTCTG
- Phenotype
-
MIM: 607360
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LACRT
PubMed Links
Gene Details
- Gene
- LACRT
- Gene Name
- lacritin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033277.1 |
119 |
Silent Mutation |
ACA,ACG |
T27T |
NP_150593.1 |
View Full Product Details