Product Details

SNP ID
rs138484168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:123292771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTCTTACTAATTTATATACACAT[A/G]TAAAGAGAAGATACTGAAGTGTCTA
Phenotype
MIM: 614274
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SBNO1 PubMed Links

Gene Details

Gene
SBNO1
Gene Name
strawberry notch homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167856.1 7357 UTR 3 NP_001161328.1
NM_018183.3 7357 UTR 3 NP_060653.3
XM_005253572.3 7357 UTR 3 XP_005253629.1
XM_005253573.4 7357 UTR 3 XP_005253630.1
XM_005253575.3 7357 UTR 3 XP_005253632.1
XM_005253576.3 7357 UTR 3 XP_005253633.1
XM_006719473.3 7357 UTR 3 XP_006719536.1
XM_006719474.3 7357 UTR 3 XP_006719537.1
XM_011538533.2 7357 UTR 3 XP_011536835.1
XM_011538534.2 7357 UTR 3 XP_011536836.1
XM_011538535.2 7357 UTR 3 XP_011536837.1
XM_017019556.1 7357 UTR 3 XP_016875045.1
XM_017019557.1 7357 UTR 3 XP_016875046.1
XM_017019558.1 7357 UTR 3 XP_016875047.1
XM_017019559.1 7357 UTR 3 XP_016875048.1

View Full Product Details