Product Details

SNP ID
rs140955399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:47965345 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCAGAGATTGCGCCCTCAGCGAA[C/G]GTGAGTTACCTGCTTCTCACCTGTT
Phenotype
MIM: 120140
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
COL2A1 PubMed Links

Gene Details

Gene
COL2A1
Gene Name
collagen type II alpha 1 chain
There are no transcripts associated with this gene.

Gene
TMEM106C
Gene Name
transmembrane protein 106C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143841.1 437 Missense Mutation ACT,AGT T84S NP_001137313.1
NM_001143842.1 437 Missense Mutation ACT,AGT T84S NP_001137314.1
NM_001143843.1 437 Missense Mutation ACT,AGT T84S NP_001137315.1
NM_024056.3 437 Missense Mutation ACT,AGT T84S NP_076961.1

View Full Product Details