Product Details

SNP ID
rs142398265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110057264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAAGCTCTGCAGGGAGGATGTGC[C/T]TTTCAGCAAAGTTCCTCTCCCCCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C12orf76 PubMed Links

Gene Details

Gene
C12orf76
Gene Name
chromosome 12 open reading frame 76
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207435.2 257 Missense Mutation AAG,AGG K75R NP_997318.1
XM_017019297.1 257 Missense Mutation AAG,AGG K86R XP_016874786.1

View Full Product Details