Product Details

SNP ID
rs144932093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:63781117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAACTAGCCTTCAAATATTAGAT[A/G]AATCCACGAAAGGAAAAACAGATCT
Phenotype
MIM: 605862
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM5 PubMed Links

Gene Details

Gene
TMEM5
Gene Name
transmembrane protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278237.1 445 UTR 5 NP_001265166.1
NM_014254.2 445 Missense Mutation AAA,GAA K90E NP_055069.1
XM_005268562.3 445 UTR 5 XP_005268619.1
XM_005268563.3 445 UTR 5 XP_005268620.1
XM_017018686.1 445 UTR 5 XP_016874175.1
XM_017018687.1 445 UTR 5 XP_016874176.1

View Full Product Details