Product Details

SNP ID
rs148238970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54282625 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTCGTGGAGGTGGTTTCGGTGG[A/G]AATGACAACTTCGGTCGTGGAGGAA
Phenotype
MIM: 604478 MIM: 164017 MIM: 601490
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CBX5 PubMed Links

Gene Details

Gene
CBX5
Gene Name
chromobox 5
There are no transcripts associated with this gene.

Gene
HNRNPA1
Gene Name
heterogeneous nuclear ribonucleoprotein A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002136.3 748 Intron NP_002127.1
NM_031157.3 748 Intron NP_112420.1
XM_005268826.1 748 Silent Mutation GGA,GGG G212G XP_005268883.1
XM_017019251.1 748 Intron XP_016874740.1
Gene
NFE2
Gene Name
nuclear factor, erythroid 2
There are no transcripts associated with this gene.

View Full Product Details