Product Details
- SNP ID
-
rs149584000
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:49128924 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAAAACCAAGAAGCCCTGAAGACC[A/G]GTGCACTGGTCAGCCTGTAGGGGAA
- Phenotype
-
MIM: 602530
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TUBA1B
PubMed Links
Gene Details
- Gene
- TUBA1B
- Gene Name
- tubulin alpha 1b
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006082.2 |
611 |
Silent Mutation |
|
|
NP_006073.2 |
View Full Product Details