Product Details

SNP ID
rs149868580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49371498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTCCACCTTTTGGCTGTTGTGA[A/G]TAATGGTGCTGTGAATTTTGGTGTA
Phenotype
MIM: 611667
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPATS2 PubMed Links

Gene Details

Gene
SPATS2
Gene Name
spermatogenesis associated serine rich 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293285.1 Intron NP_001280214.1
NM_001293286.1 Intron NP_001280215.1
NM_023071.3 Intron NP_075559.2
XM_011538690.2 Intron XP_011536992.1
XM_011538691.2 Intron XP_011536993.1
XM_017019840.1 Intron XP_016875329.1

View Full Product Details