Product Details

SNP ID
rs143780872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:94578116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGACAGTGAGAAATGCTTCTAC[A/C]ACATCAGTAGCATAAAGGAAGTTTC
Phenotype
MIM: 616146
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TGDS PubMed Links

Gene Details

Gene
TGDS
Gene Name
TDP-glucose 4,6-dehydratase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304430.1 929 Silent Mutation GTG,GTT V206V NP_001291359.1
NM_014305.3 929 Silent Mutation GTG,GTT V238V NP_055120.1
XM_011521065.2 929 Silent Mutation GTG,GTT V206V XP_011519367.1
XM_011521066.2 929 Silent Mutation GTG,GTT V206V XP_011519368.1

View Full Product Details