Product Details
- SNP ID
-
rs150312952
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:41131297 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATTCTGAGCTGGTTTATACACCCA[C/G]AGGTCTGTCCTGGGCTGAGCAGCTA
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KBTBD6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs584594] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KBTBD6
- Gene Name
- kelch repeat and BTB domain containing 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152903.4 |
1504 |
Silent Mutation |
CTC,CTG |
L405L |
NP_690867.3 |
- Gene
- LOC101929140
- Gene Name
- uncharacterized LOC101929140
There are no transcripts associated with this gene.
View Full Product Details