Product Details

SNP ID
rs139103808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24607196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGATCCCCCACACAAATCTCAG[G/T]GGCTCTGCTGTAATTGCCATGGAAG
Phenotype
MIM: 116831
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GZMH PubMed Links

Gene Details

Gene
GZMH
Gene Name
granzyme H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270780.1 657 Intron NP_001257709.1
NM_001270781.1 657 Intron NP_001257710.1
NM_033423.4 657 Missense Mutation ACT,CCT T184P NP_219491.1
XM_011536683.2 657 Missense Mutation ACT,CCT T150P XP_011534985.1

View Full Product Details