Product Details

SNP ID
rs139379867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77462161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGACATTGCATTGGTTTTGACAGG[C/T]ACTTCTAAGTCAGGAGTACAATACA
Phenotype
MIM: 608466 MIM: 613401
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AHSA1 PubMed Links

Gene Details

Gene
AHSA1
Gene Name
activator of Hsp90 ATPase activity 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321441.1 299 UTR 5 NP_001308370.1
NM_012111.2 299 Silent Mutation GGC,GGT G91G NP_036243.1
Gene
VIPAS39
Gene Name
VPS33B interacting protein, apical-basolateral polarity regulator, spe-39 homolog
There are no transcripts associated with this gene.

View Full Product Details