Product Details

SNP ID
rs140360367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24226603 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGCCTTGAATGAGATAACATTAT[A/G]TAACACTAGATATAGTGTTTTAATT
Phenotype
MIM: 610781 MIM: 603171
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GMPR2 PubMed Links

Gene Details

Gene
GMPR2
Gene Name
guanosine monophosphate reductase 2
There are no transcripts associated with this gene.

Gene
NEDD8
Gene Name
neural precursor cell expressed, developmentally down-regulated 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006156.2 Intron NP_006147.1
Gene
NEDD8-MDP1
Gene Name
NEDD8-MDP1 readthrough
There are no transcripts associated with this gene.

View Full Product Details