Product Details

SNP ID
rs142329406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:39039847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCTAGATCTCAGTTTTATTATCT[A/G]TTAAATGAAAATAACACTAGAATTC
Phenotype
MIM: 610511
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEC23A PubMed Links

Gene Details

Gene
SEC23A
Gene Name
Sec23 homolog A, coat complex II component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006364.3 Intron NP_006355.2
XM_005267262.1 Intron XP_005267319.1
XM_011536355.2 Intron XP_011534657.1
XM_017020928.1 Intron XP_016876417.1

View Full Product Details