Product Details

SNP ID
rs143577249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:64541524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCATCTTCGAGGTGAAGTCCAC[G/T]GCCGGCGACACCCACCTGGGCGGTG
Phenotype
MIM: 140560 MIM: 616578
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HSPA2 PubMed Links

Gene Details

Gene
HSPA2
Gene Name
heat shock protein family A (Hsp70) member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021979.3 1057 Silent Mutation ACG,ACT T225T NP_068814.2
Gene
LOC102723809
Gene Name
uncharacterized LOC102723809
There are no transcripts associated with this gene.

Gene
PPP1R36
Gene Name
protein phosphatase 1 regulatory subunit 36
There are no transcripts associated with this gene.

Gene
ZBTB1
Gene Name
zinc finger and BTB domain containing 1
There are no transcripts associated with this gene.

View Full Product Details