Product Details
- SNP ID
-
rs146602887
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:57471504 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTGTCCCTGGGCCTCTTGTTCAT[A/G]AAGCCATGTTTCCATTTTGCCAATT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C14orf105
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1152530] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C14orf105
- Gene Name
- chromosome 14 open reading frame 105
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001283056.1 |
1010 |
Missense Mutation |
CAT,TAT |
H288Y |
NP_001269985.1 |
NM_001283057.1 |
1010 |
Missense Mutation |
CAT,TAT |
H247Y |
NP_001269986.1 |
NM_001283058.1 |
1010 |
Missense Mutation |
CAT,TAT |
H159Y |
NP_001269987.1 |
NM_001283059.1 |
1010 |
Missense Mutation |
CAT,TAT |
H159Y |
NP_001269988.1 |
NM_001283060.1 |
1010 |
Intron |
|
|
NP_001269989.1 |
NM_018168.3 |
1010 |
Missense Mutation |
CAT,TAT |
H248Y |
NP_060638.2 |
XM_005267806.1 |
1010 |
Intron |
|
|
XP_005267863.1 |
XM_005267810.3 |
1010 |
Missense Mutation |
CAT,TAT |
H169Y |
XP_005267867.1 |
XM_005267811.3 |
1010 |
Missense Mutation |
CAT,TAT |
H160Y |
XP_005267868.1 |
XM_005267813.1 |
1010 |
Missense Mutation |
CAT,TAT |
H122Y |
XP_005267870.1 |
XM_005267814.1 |
1010 |
Missense Mutation |
CAT,TAT |
H121Y |
XP_005267871.1 |
XM_006720188.2 |
1010 |
Intron |
|
|
XP_006720251.1 |
XM_006720189.2 |
1010 |
Missense Mutation |
CAT,TAT |
H170Y |
XP_006720252.1 |
View Full Product Details