Product Details

SNP ID
rs148091928
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:34435065 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAGTCAGGATGATTTTCCTGTT[C/G]TACTCATGAATTTTAGTCTTTATAA
Phenotype
MIM: 613540
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
SPTSSA PubMed Links

Gene Details

Gene
SPTSSA
Gene Name
serine palmitoyltransferase small subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138288.3 407 UTR 3 NP_612145.2

View Full Product Details