Product Details

SNP ID
rs148125211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:93720580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCCCACAGTGGATTTGGCACAA[A/G]TAGTGGCCTCTGCTGACTAAGGCAG
Phenotype
MIM: 613851
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRIMA1 PubMed Links

Gene Details

Gene
PRIMA1
Gene Name
proline rich membrane anchor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178013.3 1368 UTR 3 NP_821092.1
XM_011536456.2 1368 UTR 3 XP_011534758.1

View Full Product Details